Learn More
Eppendorf™ Protein LoBind™ Tubes
Product Code. 17124364 Shop All Eppendorf Products
Change view
Click to view available options
Capacity (Metric):
0.5 mL
1.5 mL
15 mL
2 mL
25 mL
5 mL
50 mL
Packaging:
1 Bag x 500 Tubes
100 Tubes (2 Bags x 50 Tubes)
200 Tubes (2 Bags x 100 Tubes)
200 Tubes (4 Bags x 50 Tubes)
200 Tubes (8 Bags x 25 Tubes)
Closure Type:
Screw Cap
Snap Cap
Max. RCF:
17,000 x g
18,000 x g
19,500 x g
25,000 x g
Unit Size:
Pack of 100
Pack of 200
Pack of 500
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Packaging Closure Type Max. RCF Unit Size
17124364 0.5 mL 1 Bag x 500 Tubes Snap Cap 18,000 x g Pack of 500
17463703 25 mL 200 Tubes (4 Bags x 50 Tubes) Screw Cap 17,000 x g Pack of 200
17134364 1.5 mL 1 Bag x 500 Tubes Snap Cap 18,000 x g Pack of 500
17144364 2 mL 1 Bag x 500 Tubes Snap Cap 18,000 x g Pack of 500
15925180 15 mL 200 Tubes (4 Bags x 50 Tubes) Screw Cap 19,500 x g Pack of 200
15945180 50 mL 200 Tubes (8 Bags x 25 Tubes) Screw Cap 19,500 x g Pack of 200
15178344 5 mL 100 Tubes (2 Bags x 50 Tubes) Snap Cap 25,000 x g Pack of 100
16180371 5 mL 200 Tubes (2 Bags x 100 Tubes) Screw Cap 25,000 x g Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 17124364 Supplier Eppendorf™ Supplier No. 0030108434
In Stock
Add to basket
Edge
Add to basket
Engineered to minimize sample binding and elevate recovery, Eppendorf™ Protein LoBind™ Tubes safeguard your valuable proteins during processing. With cutting-edge LoBind technology, ensure optimal yields for your Western blotting, mass spectrometry, and protein purification applications.
Engineered to minimize sample binding and elevate recovery, Eppendorf™ Protein LoBind™ Tubes safeguard your valuable proteins during processing. With cutting-edge LoBind technology, ensure optimal yields for your Western blotting, mass spectrometry, and protein purification applications.
  • Surface-coating-free, including substances like silicone, plasticizers, biocides or mold release agents to reduce the risk of sample interference.
  • Lot-certified PCR clean purity grade guarantees freedom from human DNA, DNase, RNase, and PCR inhibitors.
  • Available in tube, microplate, and deep-well plate formats for convenient upscaling of experiments.
  • Snap, screw, and SnapTec™ cap options available.
TRUSTED_SUSTAINABILITY
Specifications
Closure Material HDPE
Color Colorless
Capacity (Metric) 0.5 mL
Material Polypropylene
No. per Pack 500 Pk.
Packaging 1 Bag x 500 Tubes
Sterility Non-sterile
Autoclavable Autoclavable
Bottom Shape Conical
Closure Type Snap Cap
Graduated No
Product Type Microcentrifuge Tube (PCR Clean, Protein LoBind)
Max. RCF 18,000 x g
Purity or Quality Grade PCR Clean (Non-sterile)