Learn More
Novus Biologicals™ Protein Bop Overexpression Lysate
Product Code. 18037327 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
Product Code. 18037327
missing translation for 'mfr': Novus Biologicals™ NBP206292
The Protein Bop Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Cell Line The lysate was created in HEK293T cells, using plasmid ID RC215316 and based on accession number NP_7893. The protein contains a C-terminal DDK tag.
For Use With (Application) Western Blot
Quantity 0.1 mg
Storage Buffer RIPA buffer
Purity Protein
Test Specificity Homo sapiens C22orf29, mRNA.
Content And Storage -80°C. Avoid freeze-thaw cycles.
Protein State Native