Learn More
Cytiva Protein A
Shop All Cytiva Products
Click to view available options
Quantity:
50 mg
Unit Size:
Each
Description
Staphylococcal Protein A is the classical immunoglobulin-binding reagent for a wide range of antibodies, used extensively in affinity chromatography and antibody detection applications. The protein is enterotoxin-free and gives reliable results.
- Staphylococcal Protein A
- Protein A binds the Fc regions of IgG antibodies from a variety of species, for convenient antibody purification.
Specifications
Specifications
| Product Type | Protein A |
| Quantity | 50 mg |