Learn More
Honeywell Safety Products™ Protege Safety Glasses
Click to view available options
:
Lens Type: Impact Resistant
Description
- General use and protection against impact (45m/s).
- Well suited for applications such as utilities, light and heavy manufacturing, bio tech, ship building, construction and automotive manufacturing.
- Smart style, stylish sunglass look.
- Lightweight design.
- Spring hinge temples.
- Bendable temples for adjusting fit.
- Soft nose bridge to prevent slippage secure fit.
- High protection wrap-around 7-base polycarbonate lenses.
- Excellent coverage and protection.
Specifications
Specifications
| Certifications/Compliance | EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Certificate Number EC1789, EC Attestation Number EC1789 |
| Lens Coating | Impact-resistant, Scratch-resistant |
| Product Type | Safety Glasses |
| For Use With (Application) | Administration, Agriculture, Automotive, Aviation, Building, Construction, Catering, Fishing, Food, Iron, Steel, Laboratory, Logistics, Maintenance, Medical, Pharmaceutical, Paper, Printing, Service, Ship Building, Telecom, Textile, Transportation, Utility, Water Treatment and Wood industries. |
| Frame Color | Black |
| No. per Pack | 1 Ea. |
| Lens Material | Polycarbonate |
| Frame Material | Plastic |