Learn More
Labconco™ Protector™ Solvent Storage Cabinet
Product Code. 15932481 Shop All Labconco Corporation Products
Click to view available options
Door Style:
Manual-close, Double
Self-close, Double
No. of Doors:
2
Width (English) Exterior:
36 in.
48 in.
Unit Size:
Each
Product Code. 15932481
Brand: Labconco™ 9902000
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Safely store, vent solvents and other flammable liquids with the Protector™ Solvent Storage Cabinet.
Safely store, vent solvents and other flammable liquids with the Protector™Solvent Storage Cabinet.
  • Key-locked doors are manual closing or self-closing/self-latching with sequencing mechanism
  • Self-closing door(s) close automatically in case of fire when temperature reaches 74°C (165°F)
  • Powder-coated steel interior shelf with five height settings
Specifications
Depth (Metric) Exterior 76 cm
Description Glacier White; Liner: Polyethylene
Dimensions (D x W x H) 22 x 48 x 35.5 to 36.75 in.
Door Style Manual-close, Double
For Use With (Application) Storing flammables
Height (Metric) Exterior 90.17 to 93.3 cm
Includes Two rare vent connections with flame arrestors and closure plugs, 4 leveling feet, 8 in. filler panel, vacuum-formed PVC liner tray, Epoxy Coated Steel shelf
Load Bearing Capacity Up to 800 lb.
No. of Doors 2
No. of Shelves 1
Shelf Type Epoxy-coated Steel
Weight (English) 260 lb.
Weight (Metric) 118 kg
Width (Metric) Exterior 121.92 cm
Color Light Gray
Depth (English) Exterior 29.9 in.
Height (English) Exterior 35.5 to 36.75 in.
Height (Metric) Opening 62.23 cm.
Material Steel
Product Type Solvent Storage Cabinet
Width (English) Exterior 48 in.