Learn More
3M™ Protective Coverall 4530
Product Code. p-200003846
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Unit Size:
Pack of 20
Product Code. 17815676
Brand: 3M™ 7000109011
Only null left
Add to basket
Add to basket
Protective Coverall 4530 has a specialist material treatment which offers fabric flame spread resistance to allow the garment to be worn over an Index 2/3 FR garment, enhancing the chemical splash and hazardous dust protection and extending the life cycle through reduced laundering.
  • Use 3M™ Protective Coverall 4530 for protection against dirt, splashes and toxic dusts, including nuclear particles.
  • It meets a variety of standards for chemical protective clothing including EN 1149 Antistatic protection, EN 1073-2 Nuclear particulate resistance, EN ISO 13688, Type 6 Liquid Splash EN 13034 and Type 5 Dust EN ISO 13982-1.
  • You can identify it easily with the white triangle in the blue suit which is made of the same 54 gsm SMMS as the rest.
  • This coverall includes a specialist treatment of the material only (not components) which offers flame spread resistance complying with ISO 14116 Index 1.
  • This means the disposable coverall can be used with an FR Index 2/3 garment to offer improved certain chemical splash and hazardous dust performance and extend the life of the reusable garment by minimizing the need to launder.
  • The 3M™ protective coverall 4530 is soft and breathable yet robust and strong weighing in at 54 gsm.
  • It's made with SMMS which is a layer of spunbond on the inner and outer and 2 layers of meltblown material in the middle.
  • The Spunbond acts as the smooth strength and splash protection, while the Meltblown layers provide an intricate multilayered web of microfibres that filter out dry particulates.
  • Soft, breathable and very lightweight, the non-woven material allows for easier release of body heat and moisture, giving extended comfort wear for increased productivity in the workplace.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Protective Coveralls
Size Medium
Gender Unisex
Material SMS Polypropylene
Color Blue
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Knit
Neck Style Hooded
Hood Type 3-panel
Closure Type 2-way Zipper with Storm Flap
Seam Type Serged
Sterility Non-sterile
Protection Class Category III
Resistance Flame, Chemicals, Liquid, Anti-static
Certifications/Compliance Nuclear Particulate Resistance (EN 1073-2), Type 5 Dust (EN ISO 13982-1), Type 6 Liquid Splash (EN 13034), Flame-Spread Resistant (ISO 14116 Index 1), and Antistatic (EN 1149)
No. of Pockets None
Width (English) Chest 36.2 to 39.4 in.
Width (Metric) Chest 92 to 100 cm
Height (English) 65.7 to 69.3 in.
Height (Metric) 167 to 176 cm
Weight (Metric) 54 gsm