Learn More
Uvex™ Protection Flash Hi-Viz Yellow Vest
Product Code. p-7228502 Shop All Uvex Products
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Each
Product Code. 17737529
Brand: Uvex™ 1732310
Only null left
Add to basket
Add to basket
The Uvex protection flash collection uses bright reflective strips for visibility in rain, fog, and darkness.
  • Vertical light guide system
  • Hook-and-loop fastening to quickly close the vest
  • Certified to EN ISO 20471 Class 2
  • For use with light guide system: battery pack, art. no. 1733200
  • Active warning
  • With switch for easy operation
  • Fast closure
  • Easy to use
  • Long run-time
  • Areas of application: road construction works, structural and civil engineering, in dark environments
  • Washing instructions: 40°C gentle cycle, do not use chlorine, do not tumble dry, do not iron, dry clean with perchlorethylene
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN ISO 20471, Class 2
Closure Type Zipper
Product Type High-visibility Vest
Color Yellow
Size Medium
Material Polyester