missing translation for 'onlineSavingsMsg'
Learn More

Uvex™ Protection Flash Hi-Viz Yellow Vest

Product Code. 17767529 Shop All Uvex Products
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17767529

Brand: Uvex™ 1732313

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

The Uvex protection flash collection uses bright reflective strips for visibility in rain, fog, and darkness.

  • Vertical light guide system
  • Hook-and-loop fastening to quickly close the vest
  • Certified to EN ISO 20471 Class 2
  • For use with light guide system: battery pack, art. no. 1733200
  • Active warning
  • With switch for easy operation
  • Fast closure
  • Easy to use
  • Long run-time
  • Areas of application: road construction works, structural and civil engineering, in dark environments
  • Washing instructions: 40°C gentle cycle, do not use chlorine, do not tumble dry, do not iron, dry clean with perchlorethylene
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN ISO 20471, Class 2
Closure Type Zipper
Product Type High-visibility Vest
Color Yellow
Size 2X-Large
Material Polyester
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.