missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ proTAME
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
ProTAME is a cell-permeable APC/C (anaphase-promoting complex/cyclosome) inhibitor. It is converted to TAME (Tosyl-L-Arginine Methyl Ester) by intracellular esterases. It induces cell cycle arrest in metaphase followed by cell death.
Specifications
Specifications
| Chemical Name or Material | Methyl (2S)-2-[[(4-methylphenyl)sulfonyl]amino]-9,13-dioxo-14-phenyl-7-[[[[(2-phenylacetyl)oxy]methoxy]carbonyl]amino]-10,12-dioxa-6,8-diazatetradec-6-enoate |
| CAS | 1362911-19-0 |
| Quantity | 1 mg |
| Target | APC |
| Molecular Formula | C34H38N4O12S |
| Purity | 90% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction