Learn More
Novus Biologicals™ Prostate Tissue Slides (Adult Normal)- Paraffin
Product Code. 18118153 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
5 Slides
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18118153 5 Slides Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Product Code. 18118153 Supplier Novus Biologicals™ Supplier No. NBP267986
  • ONLINE EXCLUSIVE PRICE
    330.00€
    311.85€ / Pack of 5
    Save 18.15€
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 01-10-2026
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
This product is for research use only and is not approved for use in humans or in clinical diagnosis.
The Human Prostate Tissue Lysate (Adult Normal) is derived from human.
TRUSTED_SUSTAINABILITY
Specifications
Tissue Brain
Quantity 5 Slides
Storage Buffer HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
Tissue Condition Normal
Cross Reactivity Fas Receptor/TNFRSF6/CD95
Host Species Human
Content And Storage Store at -80°C. Avoid freeze-thaw cycles.
Life Stage Adult