Learn More
DuPont™ Proshield™ 4 Practik
Product Code. 16263512 Shop All DuPont Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Unit Size:
Pack of 25
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
16263512 Medium Pack of 25
16253512 Large Pack of 25
16283502 X-Large Pack of 25
16273502 2X-Large Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 16263512 Supplier DuPont™ Supplier No. D13495537
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Proshield™ 4 Practik suits are the ideal work suits for general maintenance in industrial manufacturing and other harmless environments.
  • Micro-perforated non-woven polyethylene fabric allows unrestricted passage of air and water vapour
  • Certified according to Regulation (EU) 2016/425
  • Chemical protective clothing, Category I
  • Antistatic treatment (EN 1149-5) on inside for comfort
  • Versatile protective garment for non-hazardous substances
TRUSTED_SUSTAINABILITY
Specifications
Size Medium
Gender Unisex
Material Polyethylene
Color White
Disposable Single-use
Cuff Style Elastic
Neck Style Hooded
Closure Type Zipper
Sterility Non-sterile
Sleeve Style Elastic