Learn More
Contec™ PROSAT™ Presaturated Nonwoven Wipes
BIOPHARMAGREENER_CHOICE
Product Code. 15755533 Shop All Contec Products
Click to view available options
Dimensions (L x W):
20 x 20 cm (8 x 8 in.)
28 x 23 cm (11 x 9 in.)
Material:
Polyester Cellulose
Polypropylene
Saturation Solution:
70% IPA, 30% DI Water
Packaging Quantity:
1800 Wipes/Cs.
24 Pk., 50 Pk./Cs.
24 Wipes/Bag
50 Pk., 24 Pk./Cs.
50 Pk., 28 Pk./Cs.
50 Pk., 40 Pk./Cs.
50 Pk., 50 Pk./Cs.
50 Wipes/Bag
50 Wipes/Bag, 40 Bags/Cs.
Unit Size:
0.3kg
Pack of 1200
Pack of 1400
Pack of 2000
Pack of 2500
Pack of 50
Product Code. 15755533
Brand: Contec™ PSPP0043
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Use these wipes to clean laminar flow workbenches, cleanroom surface and equipment. PROSAT™ Presaturated Nonwoven Wipes feature task-specific saturation level of the solvent or disinfectant required to do the job.
Use these wipes to clean laminar flow workbenches, cleanroom surface and equipment. Contec™ PROSAT™ Presaturated Nonwoven Wipes feature task-specific saturation level of the solvent or disinfectant required to do the job.
  • Wipe substrate designed for the application
  • Compatible with ISO Class 5 (FED-STD-209E Class 100/M3.5) environments
  • Ideal for wiping articles prior to pass through
  • Made of meltblown polypropylene or cellulose/polyester
  • Effective and convenient for wipe-downs of process tools and other equipment
  • Choose spunlaced 100% polyester wipers for improved cleanliness, meltblown polypropylene wipes for improved durability, or economical hydroentangled polyester/cellulose wipers
  • Solvent blend meets SEMI specifications for metals and contaminants
TRUSTED_SUSTAINABILITY
Specifications
Product Type Pre-saturated Wipe
Length (English) 11 in.
Length (Metric) 28 cm
Width (English) 11 in.
Width (Metric) 23 cm
Material Polypropylene
Autoclavable Not Autoclavable
Certifications/Compliance ISO Class 5
Disposable Single-use
Dimensions (L x W) 28 x 23 cm (11 x 9 in.)
For Use With (Application) For interior surfaces of process tools, surface preparation, manufacturing processes
For Use With (Equipment) Process tools and other equipment
Packaging Quantity 50 Wipes/Bag, 40 Bags/Cs.
Ply 1-ply
Saturation Solution 70% IPA, 30% DI Water
Sterility Non-sterile
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • CONTEC PROSAT MBPP 70/30 IPA & DI WATER
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P370+P378-In case of fire: Use media other than water to extinguish.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contains : propan-2-ol