missing translation for 'onlineSavingsMsg'
Learn More

Propyl Benzenesulfonate, TRC

Product Code. 30425093
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30425093 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30425093 Supplier Toronto Research Chemicals Supplier No. TRCP835050100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Propyl Benzenesulfonate
CAS 80-42-2
Molecular Formula C9H12O3S
Quantity 100 mg
Synonym Benzenesulfonic Acid Propyl Ester, NSC 3216
InChI Formula InChI=1S/C9H12O3S/c1-2-8-12-13(10,11)9-6-4-3-5-7-9/h3-7H,2,8H2,1H3
SMILES CCCOS(=O)(=O)c1ccccc1
IUPAC Name propyl benzenesulfonate
Molecular Weight (g/mol) 200.26
Formula Weight 200.0507
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.