Learn More
Propyl 4-hydroxybenzoate, 99%, BLD Pharm™
Specifications
Specifications
| CAS | 94-13-3 |
| Molecular Formula | C10H12O3 |
| MDL Number | MFCD00002354 |
| Quantity | 1 kg |
| SMILES | O=C(OCCC)C1=CC=C(O)C=C1 |
| Molecular Weight (g/mol) | 180.20 |
| Percent Purity | 99% |
| Chemical Name or Material | Propyl 4-hydroxybenzoate |