missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propoxur-d3 (N-methyl-d3), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Propoxur D3 (N-methyl D3) |
| CAS | 1219798-56-7 |
| Molecular Formula | C11 2H3 H12 N O3 |
| Quantity | 0.05 g |
| Synonym | Propoxur D3 (N-methyl D3), Propoxur D3 (methyl D3) |
| InChI Formula | InChI=1 S/C11H15NO3/c1-8(2)14-9-6-4-5-7-10(9)15-11(13)12-3/h4-8 H,1-3H3,(H,12,13)/i3D3 |
| SMILES | [2 H]C([2 H])([2 H])NC(=O)Oc1ccccc1OC(C)C |
| IUPAC Name | (2-propan-2-yloxyphenyl) N-(trideuteriomethyl)carbamate |
| Molecular Weight (g/mol) | 212.26 |
| Formula Weight | 212.124 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction