Learn More
Propiophenone-d10, CDN
Specifications
Specifications
| Chemical Name or Material | Propiophenone-D10 |
| CAS | 288400-96-4 |
| Molecular Formula | C9 D10 O |
| Quantity | 0.5 g |
| Synonym | 1-Propanone-2,2,3,3,3-d5, 1-(phenyl-d5)- (9 CI), Propiophenone-d10, Propiophenone-D10, 1-(Phenyl-d5)-1-propanone-2,2,3,3,3-d5 |
| InChI Formula | InChI=1 S/C9H10O/c1-2-9(10)8-6-4-3-5-7-8/h3-7 H,2H2,1H3/i1D3,2D2,3 D,4 D,5 D,6 D,7 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C(=O)C([2 H])([2 H])C([2 H])([2 H])[2 H])c([2 H])c1[2 H] |
| IUPAC Name | 2,2,3,3,3-pentadeuterio-1-(2,3,4,5,6-pentadeuteriophenyl)propan-1-one |
| Molecular Weight (g/mol) | 144.237 |
| Formula Weight | 144.136 g/mol |
| Show More |