missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propionic-d5 Acid, CDN
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Specifications
Specifications
| Chemical Name or Material | Propionic Acid-d5 |
| CAS | 60153-92-6 |
| Molecular Formula | C3 D5 H O2 |
| Quantity | 1 g |
| Synonym | Propanoic-d5 acid (9 CI), Propionic acid D5, Propionic acid-d5, Propanoic-d5 acid, Propanoic-2,2,3,3,3-d5 acid |
| InChI Formula | InChI=1 S/C3H6O2/c1-2-3(4)5/h2H2,1H3,(H,4,5)/i1D3,2D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C(=O)O |
| IUPAC Name | 2,2,3,3,3-pentadeuteriopropanoic acid |
| Molecular Weight (g/mol) | 79.109 |
| Formula Weight | 79.068 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction