Learn More
Propionic Acid-d6, CDN
Product Code. 30319335
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30319335 1 g 1g
30312322 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30319335 Supplier CDN Supplier No. CDND02861G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Propionic acid D6
CAS 19448-61-4
Molecular Formula C3 D6 O2
Quantity 1 g
Synonym Propionic-d5 acid-d (8 CI), Propionic acid (C2D5COOD), Propionic acid-d6, Propanoic-d5 acid-d (9 CI)
InChI Formula InChI=1 S/C3H6O2/c1-2-3(4)5/h2H2,1H3,(H,4,5)/i1D3,2D2/hD
SMILES [2 H]OC(=O)C([2 H])([2 H])C([2 H])([2 H])[2 H]
IUPAC Name deuterio 2,2,3,3,3-pentadeuteriopropanoate
Molecular Weight (g/mol) 80.1155
Formula Weight 80.0744 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity