Learn More
Propane-1-sulfonamide (~90%), TRC
Specifications
Specifications
| Chemical Name or Material | Propane-1-sulfonamide |
| CAS | 24243-71-8 |
| Molecular Formula | C3H9NO2S |
| Quantity | 1 g |
| Synonym | Propane-1-sulfonamide |
| InChI Formula | InChI=1S/C3H9NO2S/c1-2-3-7(4,5)6/h2-3H2,1H3,(H2,4,5,6) |
| SMILES | CCCS(=O)(=O)N |
| IUPAC Name | propane-1-sulfonamide |
| Molecular Weight (g/mol) | 123.17 |
| Formula Weight | 123.0354 |