Learn More
Thermo Scientific™ ProPac™ IMAC-10 HPLC Columns
Description
Resolve target proteins using a single column in a high-resolution gradient run with Thermo Scientific™ ProPac™ IMAC-10 LC Columns. These analytical and semi-preparative HPLC columns provides high-purity separations of metal-binding proteins and high loading capacity for protein purification applications. The use of small, nonporous resin particles results in narrow, high-efficiency peaks, with minimal band broadening due to dispersion. Use loading columns to charge the ProPac IMAC-10 LC columns but avoid contact of the metal solution with the system.
- Retention control by imidazole concentration or pH gradient
- Stable between pH 2 to 12, compatible with common protein purification reagents (nonionic detergents, reducing agents)
- Effective under native or denaturing HPLC conditions
- Wide range of metal-specific applications, including the separation of His-tagged protein aggregation variants, prion peptide analysis and separation of monoclonal antibodies
- Shipped free from any metal and ready to be charged with the metal of your choice.
- Reusable columns with metal tailored specificity
- Resolve target proteins using a single column in a high resolution gradient run
- High purity separations of metal-binding proteins
Specifications
Specifications
| Column Format | Analytical Column |
| Diameter (Metric) | 9 mm |
| Flow Rate | 1 mL/min |
| For Use With | UltiMate 3000 HPLC System |
| Length (Metric) | 250 mm |
| Max. Pressure | 3000 psi (207 bar) |
| Mobile Phase Compatibility | pH 2 to 12, compatible with most reagents used in IMAC protein purifications, including EtOH, Urea, NaCl, non-ionic detergents, glycerol, acetic acid, guanidine, HCl |
| Particle Size | 10 μm |
| pH | 2 to 12 |
| Pore Size | Nonporous |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.