missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ProPac™ IMAC-10 HPLC Columns

Product Code. 11331823
Click to view available options
Length (Metric):
50 mm
250 mm
Diameter (Metric):
1 mm
2 mm
4 mm
9 mm
Column Format:
Analytical Column
Loading Column
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11331823

Brand: Thermo Scientific™ 063272

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Resolve target proteins using a single column in a high-resolution gradient run with ProPac IMAC-10 LC Columns.

Resolve target proteins using a single column in a high-resolution gradient run with Thermo Scientific™ ProPac™ IMAC-10 LC Columns. These analytical and semi-preparative HPLC columns provides high-purity separations of metal-binding proteins and high loading capacity for protein purification applications. The use of small, nonporous resin particles results in narrow, high-efficiency peaks, with minimal band broadening due to dispersion. Use loading columns to charge the ProPac IMAC-10 LC columns but avoid contact of the metal solution with the system.

Tailored Specificity for IMAC Separation of Proteins
  • Retention control by imidazole concentration or pH gradient
  • Stable between pH 2 to 12, compatible with common protein purification reagents (nonionic detergents, reducing agents)
  • Effective under native or denaturing HPLC conditions
  • Wide range of metal-specific applications, including the separation of His-tagged protein aggregation variants, prion peptide analysis and separation of monoclonal antibodies
  • Shipped free from any metal and ready to be charged with the metal of your choice.
  • Reusable columns with metal tailored specificity
  • Resolve target proteins using a single column in a high resolution gradient run
  • High purity separations of metal-binding proteins
Includes: 10μm, nonporous, polymeric beads coated with a hydrophilic layer, then grafted with poly(IDA) chains.
TRUSTED_SUSTAINABILITY

Specifications

Column Format Analytical Column
Diameter (Metric) 2 mm
Flow Rate 1 mL/min
For Use With UltiMate 3000 HPLC System
Length (Metric) 50 mm
Max. Pressure 3000 psi (207 bar)
Mobile Phase Compatibility pH 2 to 12, compatible with most reagents used in IMAC protein purifications, including EtOH, Urea, NaCl, non-ionic detergents, glycerol, acetic acid, guanidine, HCl
Particle Size 10 μm
pH 2 to 12
Pore Size Nonporous
Temperature 60°C
Stationary Phase Lysozyne Poly(IDA) Grafts
Product Line ProPac
Packing Material Spherical, Nonporous Polymer
Column Type Separation of Metal-Binding Proteins
Phase Affinity Separation
Type Analytical Column
Green Features Responsibly packaged
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.