Learn More
WTW™ ProfiLine™ pH 3210 Portable pH Meter
Specifications
Specifications
| Includes | Short instruction manual, CD-ROM and batteries |
| Accuracy (Temperature) | ±0.1°C |
| Accuracy (mV) | ±0.3, ±1 |
| Accuracy (pH) | ±0.1, ±0.01, ±0.005 |
| For Use With (Application) | For challenging measurements in the field and plant, e.g. water/wastewater analysis, chemical/pharmaceutical industry, etc |
| Range (pH) | -2 to 19.999 |
| Resolution (pH) | ±0.005 |
| Range (mV) | -1200 to 1200, -2500 to 2500 |
| Resolution (mV) | ±0.3, ±1mV |
| Calibration Points | 1, 2, 3, 4, 5 Calibration WTW Technical Buffer, DIN/NIST buffers plus 20 additional buffer set |
| Show More |