Learn More
Productainer™ 3D BioProcess Container, BioTitan retainers, three ports, MPCs and Luer locks
Shop All Thermo Scientific Products
Click to view available options
Capacity (Metric):
50 L
100 L
200 L
Unit Size:
Pack of 10
Pack of 5
Specifications
Specifications
| Capacity (Metric) | 50 L |
| For Use With (Application) | Single-Use Bioprocessing |
| Quantity | 50 L |
| Tubing Style | TPE |