man
Test
Search
Order Status
Quick Order
Sign In
chevron-down
chevron-down
Learn More
Home
Products
Mixers
Blenders
Standard Blenders
11371424
Print
Stomacher™ Blender 3500 laboratory 1000 to 3500mL 230V 50Hz 120W Stomacher
Brand:
Stomacher™ 3500/000/EU
Additional Details :
Weight :
0.10000kg
Product Code.
11371424
7940.00€
/ Each
Quantity
Estimated Shipment: 28-06-2024
Log in
Certificates
Accessories and Spare Parts