Learn More
Velp Scientifica™ Product Manual
Product Code. p-8012692 Shop All Velp Products
Click to view available options
For Use With (Equipment):
COEX
CSF6
DKL Series
FIWE Series Raw Fiber Extractors
GDE
HU6
JP
NDA 701 and 702
OXITEST
SER 148 Series Solvent Extractors
SMS
Unit Size:
Each
Product Code. 15309584
Brand: Velp Scientifica™ A00000192
This item requires a price update prior to being ordered.
Request a Price for this item here.
Velp Scientifica™ Product Manual are for use with a wide range of products.
  • Inlcudes one manual for the desiered product
  • For use with a wide range of Velp Scientifica™ products
Specifications
Format Manual
For Use With (Equipment) NDA 701 and 702
Product Type Operating Instructions