missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ ProcartaPlex Human TNF Superfamily Panel (15 plex)
Click to view available options
Quantity:
96 Tests
Unit Size:
96 tests
Description
Includes
- Human Standard Mix I
- Human Standard Mix K
- Human TNF Superfamily Bead Mix
- Human TNF Superfamily Detection Antibody Mix
- Reading Buffer
- Wash Buffer 10x
- Streptavidin-PE
- Universal Assay Buffer
- Detection Antibody Diluent
- PCR 8-Tube Strip
- Adhesive Film
- Greiner Microplate black 96 well
- Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma
- ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins
- This kit cannot be combined with other ProcartaPlex Simplex or Multiplex Kits
Specifications
Specifications
| Quantity | 96 Tests |
| Species | Human |
| For Use With (Application) | Multiplex Immunoassay |
| Protein Subtype | APRIL, CD137 Ligand (4-1BB Ligand), CD154 (CD40 Ligand), CD178 (Fas Ligand), CD253 (TRAIL), CD254 (RANK Ligand), CD255 (TWEAK), CD257 (BAFF, BLyS), CD258 (LIGHT), CD30L, EDA-1, GITRL, NGF beta, TNF alpha, TNF beta |
| Sample Type | Cell Culture Supernatant, Plasma, Serum |
| Sample Volume | Serum, Plasma: 25 μL; CCS: 50 μL |
| Synonym | Convenience Panel |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction