Learn More
Velp Scientifica™ Probe Extension Cable
Shop All Velp Products
Click to view available options
Length (Metric):
1 mm
Unit Size:
Each
Description
- For use with VTF Digital Thermoregulator VTF
- For use with VTF EVO Digital Thermoregulator
Specifications
Specifications
| For Use With (Equipment) | Digital Thermoregulator VTF and VTF EVO |
| Length (Metric) | 1 mm |
| Size | 39.4 in. |
| Type | Probe |