Learn More
Chemsplash Pro63 C/All with Feet Attached
GREENER_CHOICE
Product Code. p-200105906
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 25
Product Code. 30058444
Brand: Chemsplash 2607.400.004
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Pro 63 Coverall with Feet, is made from a strong yet breathable 63 gsm microporous laminated fabric.
Chemsplash Pro 63 Coverall with Feet
  • 63GSM Thermal Weld Microporous Laminate Fabric
  • Feet Attached, with Adhesive Anti Slip Sole
  • Conforms to ENISO14644-1 Class 5
  • Elasticated 3 Pce Hood
  • Low Linting Knitted Cuffs
  • Two Way Zip
  • Adhesive Zip Flap
  • Internal Hang Loop
  • Silicone and Latex Free
  • Non Linting Anti-Static Fabric
Description: The Chemsplash Pro 63 Coverall with Feet is made from a strong yet breathable 63gsm microporous laminated fabric. It is a high-quality and robust Cat III Type 5B/6B suit which meets the most stringent requirements for a variety of industries. Chemsplash Pro fabric is non-linting and anti-static to EN 1149-5 2008, making it ideal for use in wide-ranging applications where the operating environment must not be contaminated with garment fibers. For comfort and ease of fit, a knitted cuff and two-way zip have been added, and the zip also has an adhesive cover flap for extra sealing. This is our premium Cat III Type 5 and 6 coverall.
Specifications
Product Type Cleanroom Coveralls
Size Small
Gender Unisex
Material Microporous Composite Fabric
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Attached Boots
Cuff Style Knit
Neck Style Hooded
Hood Type 3-panel
Closure Type Ties
Seam Type Stitched
Sterility Non-sterile
Resistance Liquid
Latex Free Yes
No. of Pockets None
Cleanroom Class Class 100-100,000 (ISO 5-8)