Learn More
Pro-Hyp-OH, TRC
Product Code. 30008332
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008332 10 mg 10mg
30006376 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30008332 Supplier Toronto Research Chemicals Supplier No. TRCP83937510MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 18684-24-7
Synonym 4-Hydroxy-1-L-prolyl-proline∣4-Hydroxy-1-L-prolyl-L-proline∣L-Prolyl-L-hydroxyproline∣Prolylhydroxyproline
Specifications
Format Neat
Molecular Formula C10H16N2O4
MDL Number MFCD00037349
Molecular Weight (g/mol) 228.25
InChI Key ONPXCLZMBSJLSP-CSMHCCOUSA-N
SMILES O[C@@H]1C[C@H](N(C1)C(=O)[C@@H]1CCCN1)C(O)=O
IUPAC Name (2S,4R)-4-hydroxy-1-[(2S)-pyrrolidine-2-carbonyl]pyrrolidine-2-carboxylic acid
Quantity 10 mg
Shipping Condition Room Temperature