missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Olympus™ Prism, U-DICTHR
Click to view available options
For Use With (Equipment):
Biological microscopes.
Unit Size:
Each
Description
- DIC prism.
- Improves clarity and contrast for the following:
- Thick specimens.
- Layered specimens.
- High-contrast specimens.
- Color-coded: Blue
Specifications
Specifications
| Color | Blue |
| Description | For visualizing thick, structured, high-contrast specimens. |
| For Use With (Application) | Visualizing thick, structured, high-contrast specimens. |
| For Use With (Equipment) | Biological microscopes. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction