Learn More
Olympus™ Prism, U-DICT
Click to view available options
For Use With (Equipment):
Biological microscopes.
Unit Size:
Each
Description
- DIC prism.
- Improves clarity and contrast for the following:
- General specimens.
- Tissue slices.
- Color-coded: White
Specifications
Specifications
| Color | White |
| Description | For visualizing general specimens, tissue slices. |
| For Use With (Application) | Visualizing general specimens, tissue slices. |
| For Use With (Equipment) | Biological microscopes. |