Learn More
Epredia™ PrintMate™ Slotted Cassettes
Product Code. 16324036 Shop All Epredia Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Color:
Aqua
Beige
Blue
Gray
Green
Lavender
Light Green
Lilac
Orange
Pink
Tan
White
Yellow
Quantity:
10 x 75 cassettes (750)/cs
4 x 250 cassettes (1000)/cs
missing translation for 'unitSize'
Pack of 1000
Pack of 750
Product Code. Color Quantity unitSize
16324036 Light Green 10 x 75 cassettes (750)/cs Pack of 750
15487614 Aqua - Pack of 1000
16323996 Beige 4 x 250 cassettes (1000)/cs Pack of 1000
15497614 Blue - Pack of 1000
16333996 Blue 4 x 250 cassettes (1000)/cs Pack of 1000
15407624 Green - Pack of 1000
16343996 Green 4 x 250 cassettes (1000)/cs Pack of 1000
15417624 Gray - Pack of 1000
16353996 Gray 4 x 250 cassettes (1000)/cs Pack of 1000
15427624 Lilac - Pack of 1000
16363996 Lavender 4 x 250 cassettes (1000)/cs Pack of 1000
15437624 Orange - Pack of 1000
16373996 Light Green 4 x 250 cassettes (1000)/cs Pack of 1000
15447624 Pink - Pack of 1000
16383996 Lilac 4 x 250 cassettes (1000)/cs Pack of 1000
15457624 Tan - Pack of 1000
16393996 Orange 4 x 250 cassettes (1000)/cs Pack of 1000
15467624 White - Pack of 1000
16304006 Pink 4 x 250 cassettes (1000)/cs Pack of 1000
15477624 Yellow - Pack of 1000
16314006 Tan 4 x 250 cassettes (1000)/cs Pack of 1000
16324006 White 4 x 250 cassettes (1000)/cs Pack of 1000
16334006 Yellow 4 x 250 cassettes (1000)/cs Pack of 1000
16374026 Beige 10 x 75 cassettes (750)/cs Pack of 750
16384026 Blue 10 x 75 cassettes (750)/cs Pack of 750
16394026 Green 10 x 75 cassettes (750)/cs Pack of 750
16304036 Gray 10 x 75 cassettes (750)/cs Pack of 750
16314036 Lavender 10 x 75 cassettes (750)/cs Pack of 750
16334036 Lilac 10 x 75 cassettes (750)/cs Pack of 750
16344036 Orange 10 x 75 cassettes (750)/cs Pack of 750
16354036 Pink 10 x 75 cassettes (750)/cs Pack of 750
16364036 Tan 10 x 75 cassettes (750)/cs Pack of 750
16374036 White 10 x 75 cassettes (750)/cs Pack of 750
16384036 Yellow 10 x 75 cassettes (750)/cs Pack of 750
34 missing translation for 'options'
Product Code. 16324036
missing translation for 'mfr': Epredia™ B851120LG
Epredia™ PrintMate Cassettes™ are designed to contain incised tissue safely and securely for tissue processing, embedding and sectioning. PrintMate Cassettes provide exceptional scanning accuracy and reliability when used with the Epredia™ PrintMate™ and Epredia™ SlideMate™ barcode scanner.
Designed for performance and user convenience, PrintMate Slotted Cassettes have been optimized for superior workflow and printing with the PrintMate Classic and PrintMate AS printing systems. The even, textured printing face provides the optimal surface for high-quality printing every time. For additional information, the right and left sides are textured for clear handwriting with suitable pens. The lid catches have been adjusted for ease of use with low opening and closing force, while still giving the security needed while processing.Available in tubes for automatic printing or loose for manual use, and in twelve colors for enhanced workflow identification.
TRUSTED_SUSTAINABILITY
Specifications
Pore Style Slots
Closure Included Separate lids included
Quantity 10 x 75 cassettes (750)/cs
Closure Type Reversible
PrintMate Compatible Yes
Color Light Green
Product Type Slotted Cassette