Learn More
Epredia™ PrintMate™ Embedding Cassettes
Tienda Epredia Productos
missing translation for 'orderingAttributeHoverText'
Color:
Aqua
Beige
Blue
Gray
Green
Lavender
Light Green
Lilac
Orange
Pink
Tan
White
Yellow
Quantity:
10 x 75 cassettes (750)/cs
1000
4 x 250 cassettes (1000)/cs
Closure Included:
No lids
Separate lids included
missing translation for 'unitSize'
Pack of 1000
Pack of 750
Descripción
Designed for performance and user convenience, PrintMate Embedding Cassettes have been optimized for superior workflow and printing with the PrintMate Classic and PrintMate AS printing systems. The even, textured printing face provides the optimal surface for high-quality printing every time. For additional information, the right and left sides are textured for clear handwriting with suitable pens. The lid catches have been adjusted for ease of use with low opening and closing force, while still giving the security needed while processing.Available in tubes for automatic printing or loose for manual use, and in twelve colors for enhanced workflow identification.PrintMate Embedding Cassettes are also available without lids, for economical use with reuseable metal lids.
Especificaciones
Especificaciones
| Pore Style | Round Holes |
| Closure Included | No lids |
| Quantity | 4 x 250 cassettes (1000)/cs |
| Closure Type | Reversible |
| PrintMate Compatible | Yes |
| Color | Blue |
| For Use With (Equipment) | For Use with Metal Lids |
| Product Type | Embedding Cassette |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido