Learn More
JP Selecta™ Printer
Product Code. 12092185
Change view
Click to view available options
For Use With (Equipment):
Autester ST DRY PV III Autoclave
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
12092185 Autester ST DRY PV III Autoclave Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12092185 Supplier JP Selecta™ Supplier No. 4002417
Add to basket
Add to basket
For Autester ST DRY PV III Autoclaves
  • Continuous paper roll
  • Provides print-out of temperature, pressure, time, and mode
Specifications
Description Printer
For Use With (Equipment) Autester ST DRY PV III Autoclave