Learn More
Emma Safety Footwear Primus Safety Shoes
Product Code. 15989091
Click to view available options
Size:
37
48
49
Unit Size:
1 pair
Product Code. 15989091
Brand: Emma Safety Footwear MM93986837
Only null left
Add to basket
Add to basket
Surprisingly light yet provides a strong and supportive fit. Waterproof even when fully submerged, thanks to its state-of-the-art Sympatex™ uppers. The all-weather safety shoe offers outstanding foot protection in all seasons.
  • S3-Safety shoe with sole plate for wet work
  • Full grain leather / Waterproof construction
  • Waterproof Sympatex™ uppers
  • GripForce™ Duo Heat Resistant Outsole
  • Sanitized
Specifications
Certifications/Compliance EN ISO 20345:2011
Closure Type Laces
Color Black, Gray
Design Primus
Liner Material Hydro-Tec™ Sanitized Silver / Sympatex™
Material Leather
Size 37
Toe Material Steel