Learn More
Zeiss™ Primostar 3 for Phase Contrast
Product Code. 17382800
Change view
Click to view available options
Condenser Type:
ABBE condenser 0.9
Abbe condenser 0.9/1.25
Unit Size:
1 set
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Condenser Type Unit Size
17382800 Abbe condenser 0.9/1.25 1 set
17180676 ABBE condenser 0.9 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17382800 Supplier Zeiss™ Supplier No. 4155010021000
  • ONLINE EXCLUSIVE PRICE
    3176.25€
    2592.00€ / 1 sets
    Save 584.25€
    18% Off
View all promotions
 Restricted Item
Estimated Shipment: 20-11-2026
Robust and compact upright microscope for digital teaching and routine lab work. Configured for transmitted brightfield microscopy with a range of accessories to expand the microscopy techniques available.
  • The Primostar 3 is a robust yet compact upright microscope for digital teaching and routine lab work.
  • Reliable and robust due to a study metal frame, whilst being light weight.
  • The Fixed-Köhler versions of Primostar 3 come pre-adjusted with a field of view of 20 mm. Selected objective lenses and eyepieces are already in place meaning no time is wasted in the set up.
  • Full-Köhler versions of Primostar 3 with field of view 22 mm give freedom to teach while using Köhler illumination. In pre-defined packages, a selection of objective lenses and eyepieces are already in place. Benefit from a relaxed posture for day-long work with enlarged stage drive.
  • Use the ZEISS Labscope app to connect all your students' microscopes plus WiFi cameras and create a collective learning experience.
  • Primostar 3 with its integrated 8.3 MPx HD WiFi-camera is the package of choice for digital classrooms. This camera offers versatile interfaces such as LAN, HDMI, Ethernet and USB-C 3.0. As an added bonus, integrated powering saves you from a jumble of cables.
  • Use Labscope, the imaging app from ZEISS, to display all the live images from your connected microscopes.
  • ZEN compatible .czi file format which includes all metadata and a separate annotation layer.
Specifications
Condenser Type Abbe condenser 0.9/1.25
For Use With (Application) Phase Contrast
Includes With slot