Learn More
Cole-Parmer™ ±0.25% Accuracy Compound Transmitter, 0.5 to 5.5 V Output
Product Code. 11796058 Shop All Cole-Parmer Products
Change view
Click to view available options
Accuracy:
±0.25%
Outputs:
0.5 to 5.5 V
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Accuracy Outputs Unit Size
11796058 ±0.25% 0.5 to 5.5 V Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11796058 Supplier Cole-Parmer™ Supplier No. 6807532
  • ONLINE EXCLUSIVE PRICE
    274.00€
    271.05€ / Pack of 1
    Save 2.95€
    1% Off
Estimated Shipment: 28-12-2026
Be more accurate, affordably with the Compound Transmitter.
  • Suitable for high-shock/vibration applications.
  • SS sensor attached to SS electrode forms a variable capacitor.
  • Economical, excellent accuracy, and high performance.
  • Ideal for industrial OEM equipment, engines, hydraulic systems, and compressor control.
  • Transducers with 4 to 20 mA output have red (positive power) and black (output) wires.
  • Transducers with 0.5 to 5.5 V output have red (positive power), green (output), and black/white (common) wires.
Specifications
Product Type Pressure Transducer VSK5
Environmental Rating IP65 (NEMA 4X)
Accuracy ±0.25%
Enclosure Material Stainless Steel, PBT, PET and Polycarbonate Blend
Outputs 0.5 to 5.5 V
Pressure Range -14.7 to 15 psi