missing translation for 'onlineSavingsMsg'
Learn More

Prednisolone Hemisuccinate, TRC

Product Code. 18070920
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18070920 1 g 1g
30134548 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18070920 Supplier Toronto Research Chemicals Supplier No. TRCP7037601G
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2920-86-7
Molecular Formula C25 H32 O8
Molecular Weight (g/mol) 460.52
Synonym (11beta)-21-(3-Carboxy-1-oxopropoxy)-11,17-dihydroxypregna-1,4-diene-3,20-dione, 11beta,17,21-Trihydroxypregna-1,4-diene-3,20-dione 21-(hydrogen succinate), Prednisolone 21-O-beta-carbonylpropionic acid, Prednisolone 21-hemisuccinate, Prednisolone 21-succinate, Prednisolone bisuccinate, Prednisolone hemisuccinate, Prednisolone succinate, Prednisolut, delta1-Hydrocortisone 21-hemisuccinate
IUPAC Name 4-[2-[(8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-10,13-dimethyl-3-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-yl]-2-oxoethoxy]-4-oxobutanoic acid
SMILES C[C@]12C[C@H](O)[C@H]3[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1CC[C@]2(O)C(=O)COC(=O)CCC(=O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C25H32O8/c1-23-9-7-15(26)11-14(23)3-4-16-17-8-10-25(32,24(17,2)12-18(27)22(16)23)19(28)13-33-21(31)6-5-20(29)30/h7,9,11,16-18,22,27,32H,3-6,8,10,12-13H2,1-2H3,(H,29,30)/t16-,17-,18-,22+,23-,24-,25-/m0/s1
Formula Weight 460.2097
Percent Purity >95%
Chemical Name or Material Prednisolone hemisuccinate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.