Learn More
Ludwig Schneider Precision thermometer, ASTM S66 C - ACCU-SAFE, , +75+105°C, L: 401 mm, DAkkS-cal. cert. 3 p.
Brand:  Ludwig Schneider 1202066S/04/3P
Encompass_Preferred
Precision thermometer, ASTM S66 C - ACCU-SAFE, , +75+105°C, L: 401 mm, DAkkS-cal. cert. 3 p.
Product Code. 16650586
  • 588.28€ / Each
Only null left
Estimated Shipment: 20-02-2026