Learn More
Ludwig Schneider Precision low temp. thermometer, stem form, liq.Alk, -100+100°C,L: 300 mm; DAkkS cal. cert. 3 p.
Brand:  Ludwig Schneider 1052003/04/3P
Encompass_Preferred
Precision low temp. thermometer, stem form, liq.Alk, -100+100°C,L: 300 mm; DAkkS cal. cert. 3 p.
Product Code. 16650526
  • 380.55€ / Each
Only null left
Estimated Shipment: 12-02-2026