Learn More
Fisherbrand™ Precision Hydrometers for Specific Gravity
Product Code. 11722263
Encompass_Preferred
Click to view available options
Specific Gravity Range:
0.590 to 0.660
1.200 to 1.420
1.400 to 1.620
1.600 to 1.670
1.660 to 1.730
Increments:
0.0005 sp.gr.
0.002 sp.gr.
Length (Metric):
300 mm
325 mm
Unit Size:
Each
Product Code. 11722263
Brand: Fisherbrand™ 2553DBQ
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
High-precision instruments for determination of density in fluids. Fisherbrand™ Precision Hydrometers for Specific Gravity do not include a thermometer in the body and feature a reference temperature of 60/60°F.
High-precision instruments for determination of fluid density available with a DAkkS or works calibration certificate.
  • Reference temperature: 60/60°F
  • Without thermometer – control thermometer sold separately
  • Single packed in transparent square plastic sleeve
Specifications
Type Hydrometer without Thermometer
Accuracy ±0.0005 sp/gr
Increments 0.0005 sp.gr.
Length (Metric) 325 mm
For Use With (Equipment) Control thermometer
Includes Plastic sleeve
Material Glass
For Use With (Application) Measuring specific gravity
Specific Gravity Range 0.590 to 0.660
Temperature of Standardization 60°F
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.