missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PrecisION™ hKv3.1 Recombinant Stable Cell Line
Click to view available options
Quantity:
2 Vials
Unit Size:
2 ampoules
Description
The PrecisION™ hKv3.1 Recombinant Stable Cell Line is a stable, engineered cell line for use in studying Kv3.1 ion channels, including drug candidates which affect channel function. This cell line enables assessment of ion channel function via detection of electrophysiological signals. The included cell line overexpresses Kv3.1 ion channels. Upon ligand/agonist induced channel activation, 1111100 This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
| Assay Entry | Ion Channel Activity |
| Cell Line | CHO-K1 |
| For Use With (Application) | Cell Lines |
| Quantity | 2 Vials |
| Target Entry | Kv3.1 |
| Accession Number | NM_004976 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction