missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX PrecisION™ hERG Recombinant Stable Cell Line

Product Code. 17314909
Encompass
Click to view available options
Quantity:
2 Vials
Unit Size:
2 ampoules
This item is not returnable. View return policy

Product Code. 17314909

Brand: DiscoverX CYL3039

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The PrecisION™ hERG Recombinant Stable Cell Line measures hERG ion channel activity via electrophysiological measurements. This cell line ships as two vials of cryopreserved cells.

The PrecisION™ hERG Recombinant Stable Cell Line is a stable, engineered cell line for use in studying hERG ion channels, including drug candidates which affect channel function. This cell line enables assessment of ion channel function via detection of electrophysiological signals. The included cell line overexpresses hERG ion channels. Upon ligand/agonist induced channel activation, response can be detected via automated or manual electrophysiological techniques. This stable modified cell line is provided as two vials of cryopreserved cells.

Specifications

Assay Entry Ion Channel Activity
Cell Line 293H
For Use With (Application) Cell Lines
Quantity 2 Vials
Target Entry hERG
Accession Number U04270
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.