Learn More
Schneider Ludwig™ Precision Density Hydrometer
Product Code. 10052894
Encompass_Preferred
Click to view available options
Length (Metric):
300 mm
Specific Gravity Range:
0.800 to 0.900 g/cm3
0.900 to 1.000 g/cm3
1.000 to 1.100 g/cm3
1.000 to 1.500 g/cm3
1.100 to 1.200 g/cm3
1.200 to 1.300 g/cm3
Unit Size:
Each
Product Code. 10052894
Brand: Schneider Ludwig™ 3031005
Only null left
Specifications
Quantity 1Each
Type Precision Density Hydrometer
Length (Metric) 300 mm
Specific Gravity Range 1.000 to 1.100 g/cm3
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.