missing translation for 'onlineSavingsMsg'
Learn More

Brannan Pre-calibrated Oral Clinical Thermometer

Product Code. 31810747
Change view
Click to view available options
Max. Temperature (Metric):
32°C
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Max. Temperature (Metric) unitSize
31810747 32°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31810747 Supplier Brannan Supplier No. TC009

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High accurate oral clinical thermometer supplied with fixed point calibration certificate.

  • Easy to use waterproof electronic thermometer suitable for all ages
  • Compact, accurate and durable
  • Large scale integration device
  • Fixed calibration point at 37°C
  • Non hazardous
  • Temp range 32°C to 42.9°C
  • Complete with battery instructions

Specifications

Calibration Points 37°C
Certifications/Compliance CE, ROHS, WEEE
Disposable Reusable
Length (Metric) 270 cm
Material ABS Plastic
Product Type Clinical Thermometer
Max. Temperature (Metric) 32°C
Min. Temperature (Metric) 42.9°C
Calibrated Yes
Immersion Length 270 cm
Display Digital LCD
Width (Metric) 24 cm
IP Rating IP22
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.