missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ PPP2R1A (Human) Recombinant Protein

Human PPP2R1A full-length ORF ( AAH01537, 1 a.a. - 589 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00005518-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16159441

  • 359.00€ / 10µg

Estimated Shipment: 19-08-2022
to see stock.
Add to basket

Description

Description

  • Sequence: MAAADGDDSLYPIAVLIDELRNEDVQLRLNSIKKLSTIALALGVERTRSELLPFLTDTIYDEDEVLLALAEQLGTFATLVGGPEYVHCLLPPLESLATVEETVVRDKAVESLRAISHEHSPSDLEAHFVPLVKRLAGGDWFTSRTSACGLFSVCYPRVSSAVKAELRQYFRNLCSDDTPMVRRAAASKLGEFAKVLELDNVKSEIIPMFSNLASDEQDSVRLLAVEACVNIAQLLPQEDLEALVMPTLRQAAEDKSWRVRYMVADKFTELQKAVGPEITKTDLVPAFQNLMKDCEAEVRAAASHKVKEFCENLSADCRENVIMSQILPCIKELVSDANQHVKSALASVIMGLSPILGKDNTIEHLLPLFLAQLKDECPEVRLNIISNLDCVNEVIGIRQLSQSLLPAIVELAEDAKWRVRLAIIEYMPLLAGQLGVEFFDEKLNSLCMAWLVDHVYAIREAATSNLKKLVEKFGKEWAHATIIPKVLAMSGDPNYLHRMTTLFCINVLSEVCGQDITTKHMLPTVLRMAGDPVANVRFNVAKSLQKIGPILDNSTLQSEVKPILEKLTQDQDVDVKYFAQEALTVLSLA
Specifications

Specifications

AAH01537
protein phosphatase 2 (formerly 2A), regulatory subunit A, alpha isoform
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PPP2R1A
GST
5518
Wheat germ expression system
10 μg
MGC786, PR65A
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Videos
SDS
Documents
Certificates
Special Offers

Special Offers