Learn More
R&D Systems™ PPM-3
Product Code. 30306747
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30306747 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30306747 Supplier R&D Systems™ Supplier No. 8116/5
 Restricted Item
Estimated Shipment: 30-11-2026
A Tocris product, part of R&D Systems. Potent and selective ERK5 Degrader (PROTAC™)
PPM-3 is a potent and selective ERK5 Degrader (PROTAC™) (IC50 = 62.4 nM). PPM-3 degrades ERK5 in HCT116, h1975, HepG2, MDA-MB-231, PC-3, and A375 tumor cell lines (DC50 values are 5.6, 11.5, 13.7, 22.7, 23.5 and 41.4 nM respectively). PPM-3 has no effect on tumor cell growth directly but influences tumor development by affecting the differentiation of macrophages.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (2S,4R)-1-((S)-2-(7-(4-(4-((5,11-Dimethyl-6-oxo-6,11-dihydro-5H-benzo[e]pyrimido[5,4-b][1,4]diazepin-2-yl)amino)-3-methoxyphenyl)piperazin-1-yl)heptanamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-((S)-1-(4-(4-methylthiazol-5-yl)phenyl)ethyl)pyrrolidine-2-carbo
CAS 3032388-42-1
Quantity 5 mg
Product Line Tocris
Target Active Degraders
Molecular Formula C54H69N11O6S
Purity 0.98