missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PPIB (Human) Recombinant Protein
Human PPIB full-length ORF ( AAH08848, 1 a.a. - 216 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00005479-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MLRLSERNMKVLLAAALIAGSVFFLLLPGPSAADEKKKGPKVTVKVYFDLRIGDEDVGRVIFGLFGKTVPKTVDNFVALATGEKGFGYKNSKFHRVIKDFMIQGGDFTRGDGTGGKSIYGERFPDENFKLKHYGPGWVSMANAGKDTNGSQFFITTVKTAWLDGKHVVFGKVLEGMEVVRKVESTKTDSRDKPLKDVIIADCGKIEVEKPFAIAKE
Specifications
AAH08848 | |
peptidylprolyl isomerase B (cyclophilin B) | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PPIB | |
GST |
5479 | |
Wheat germ expression system | |
25 μg | |
CYP-S1, CYPB, MGC14109, MGC2224, SCYLP | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |