Learn More
Hirschmann™ Polypropylene Powder Funnels
Product Code. 13158149
Click to view available options
Diameter (Metric)Top:
60mm
80mm
100mm
120mm
150mm
Unit Size:
Pack of 10
Pack of 5
Product Code. 13158149
Brand: Hirschmann™ 7760102
Ideal for use with powders and other large particles
  • Transparent
Specifications
Diameter (Metric)Top 80mm
Material Polypropylene
Height (Metric) 75 mm
Length (Metric) Stem 25mm
Quantity 10/Pk.