Learn More
Hirschmann™ Polypropylene Powder Funnels
Click to view available options
Diameter (Metric)Top:
60mm
80mm
100mm
120mm
150mm
Unit Size:
Pack of 10
Pack of 5
Description
- Transparent
Specifications
Specifications
| Diameter (Metric)Top | 120mm |
| Material | Polypropylene |
| Height (Metric) | 110 mm |
| Length (Metric) Stem | 20mm |
| Quantity | 10/Pk. |